Order Entry
ContactUsLinkComponent
LUNA RTQPCR 4X MIX NO ROX 500
Luna Probe One-Step RT-qPCR 4X Mix with UDG (No ROX), New England Biolabs
Catalog #: CA76538-640
CAS Number:  
Luna Probe One-Step RT-qPCR 4X Mix with UDG (No ROX), New England Biolabs
Catalog #: CA76538-640
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Luna probe one-step RT-qPCR 4X mix with UDG (No ROX)
  • No. of Reactions:
    500
  • Details:
    4X concentration

Specifications

About this item

The Luna Probe One-Step RT-qPCR Mix with UDG (No ROX) is supplied at a 4X concentration and enables higher amounts of sample input, which is relevant for applications where RNA present in low abundance is of interest, such as pathogen detection.

  • Simplify your reaction setup with a single-tube, 4X master mix format
  • Maximize your throughput by multiplexing up to 5 targets
  • Reduce risk of carryover contamination with thermolabile UDG and dUTP included in the mix
  • Eliminate pipetting errors with non-interfering, visible blue tracking dye
  • For instruments that do not require the ROX passive reference dye

Mix consolidates the components for one-step RT-qPCR into a single tube, including Luna WarmStart RT, Hot Start Taq DNA Polymerase, dNTPs, and Murine RNase Inhibitor in an optimized buffer. Combining Hot Start Taq DNA Polymerase with a novel, WarmStart-activated reverse transcriptase allows for dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. This formulation contains no reference dye and is compatible with any instrument that does not require ROX (if ROX normalization is needed, ROX can be added.)

Customers Who Bought This Also Bought