FEP CHEM RES TUBE 1/8 IDX13/64 OD 25FT
Catalog #: 76542-974
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought
Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
$44.53 to $296.52
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$386.44 to $1,545.78
Bard-Parker® Protected Disposable Scalpels, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
$68.14 to $587.35
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$233.35 to $2,174.33
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Viscosity Standard, Specpure®, Nominal Values at 40 and 100 °C, Thermo Scientific
Supplier: Thermo Scientific Chemicals
$206.68