FKM CHEM RIST TUBE 5/32 IDX25/64 OD 50FT
Catalog #: 76542-850
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
178,29 $