CRYOMARKERS SET OF 4
Catalog #: 76542-232
Supplier: CUSTOM BIOGENIC SYSTEMS, INC
Customers Who Bought This Also Bought
Potassium dihydrogen phosphate ≥99.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
1 033,55 $ to 4 052,62 $
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
154,66 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $