Order Entry
ContactUsLinkComponent
PLASMID MINI KIT I Q-SPIN CAPLESS
E.Z.N.A.® Plasmid DNA Mini Kit I, (Q-spin, capless)
Catalog #: CA76539-234
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Plasmid DNA Mini Kit I, (Q-spin, capless)
Catalog #: CA76539-234
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Mini plasmid kit
  • Size:
    600 Preps
  • Cat. no.:
    CA76539-234

Specifications

About this item

Isolate up to 25 µg plasmid DNA from 1 to 5 ml bacterial cultures using mini spin columns.

  • Rapid - Purification of plasmid DNA in less than 30 minutes
  • Safe - No Phenol/chloroform extractions
  • Versatile - Spin and vacuum formats available
  • High-quality - DNA is suitable for a variety of downstream applications

The E.Z.N.A.® Plasmid DNA Mini Kit I is designed to isolate up to 25 µg of high-quality plasmid DNA from 1 to 5ml bacterial cultures in less than 30 minutes. Plasmid DNA purification follows the alkaline-lysis method and is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, transformation, PCR and DNA sequencing.

Customers Who Bought This Also Bought