ContactUsLinkComponent
CAP UREA GREEN PTFE 48-400 CS1248
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Catalog #: 76539-296
Supplier:  Qorpak
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Catalog #: 76539-296
Supplier:  Qorpak
Specifications
  • Cap Size:
    48-400
  • Cat. no.:
    76539-296
Specifications
About this item
Green Thermoset Plastic Caps with F217 and PTFE liners are known for providing the widest range of chemical compatibility.
  • The color green easily identifies the F217 & PTFE liner system
  • F217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
The F217 features a low-density foam core between two LDPE discs. They are applied to the back of a PTFE liner.
Some uses for these F217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Customers Who Bought This Also Bought