Argos Technologies® Reusable Inoculating Streakers, Antylia Scientific
Catalog #: 76534-542
Supplier: Argos Technologies
Specifications
- Sterility:Non sterile
- Color:Blue
- Description:Inoculating streaker loops, small
- Material:Nichrome wire and aluminum handle
- Cat. no.:76534-542
- Inner Diameter:2.5 mm
- Gauge:26
Specifications
About this item
Nichrome wire composition cools quickly and is less expensive than platinum.
- Select, inoculate, or transfer microbiological samples
Loops are made of high quality twisted nichrome wire and attached to a light-weight aluminum handle. Handles are insulated to protect the hand during use. Loops have been designed to slide along agar surfaces without cutting or tearing.
Loops and Handles cool rapidly after sterilization and are offered calibrated (1 and 10 µl) or non-calibrated. Non-calibrated Loops and Handles are available as small, medium or large and are color coded for easy identification.
Customers Who Bought This Also Bought