Order Entry
ContactUsLinkComponent
ERLENMEYER FLASK PLAIN 90MM 250ML CS12
Thermo Scientific™ Sterile Single-Use Erlenmeyer Flasks with Plain Bottom, Thermo Scientific
Catalog #: CA76533-872
Thermo Scientific™ Sterile Single-Use Erlenmeyer Flasks with Plain Bottom, Thermo Scientific
Catalog #: CA76533-872

Specifications

  • Capacity:
    250 ml
  • O.D.:
    90 mm
  • Sterility:
    Sterile
  • Cat. no.:
    CA76533-872

Specifications

About this item

The Thermo Scientific™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom are ideal for suspension of cell culture, media preparation, mixing and storage. Made of crystal clear PETG, these flasks are robust and break resistant. Vent cap for continuous gas exchange while maintaining sterility and preventing leakage.

  • Non-pyrogenic, non-cytotoxic
  • DNase/RNase, BSE/TSE, human DNA free
  • Molded-in graduation for accurate volume assessment

Manufactured with high quality virgin resins.

Certifications: Sterilized by gamma irradiation per ISO 11137-2.
Customers Who Bought This Also Bought