Order Entry
ContactUsLinkComponent
BAG REUSABLE SILICA GEL DESICCANT 500G
SP Bel-Art Reusable Silica Gel Desiccant Bags, Bel-Art Products, a part of SP
Catalog #: 76533-640
BAG REUSABLE SILICA GEL DESICCANT 500G
SP Bel-Art Reusable Silica Gel Desiccant Bags, Bel-Art Products, a part of SP
Catalog #: 76533-640

Specifications

  • Size:
    500 g (1.0 lbs.)
  • Cat. no.:
    76533-640
  • L×W×H:
    15×5×21 cm (6×2×8.25")

Specifications

About this item

Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.

  • Durable microfiber bag securely contains the beads eliminating the risk of leakage and is unaffected by moisture
  • Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed
  • Saturated gel can be regenerated by microwaving the bag for 6 minutes
  • Can be regenerated repeatedly without degradation
Packaging: Supplied individually sealed in a moisture-proof pouch.
Customers Who Bought This Also Bought