AIR MOTOR
Catalog #: MFLX00424-NE
Supplier: MICROPUMP INC
Customers Who Bought This Also Bought
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$170.25 to $676.42
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
$186.99