GEAR PUMPHEAD
Catalog #: MFLX00404-NG
Supplier: MICROPUMP INC
Customers Who Bought This Also Bought
Quality Economy Brand Luer Lock Syringes with Attached Needle, Air-Tite
Supplier: Air-Tite
$35.02 to $279.99
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80
Sulfate, IC Standard SO42-, 1000ppm, Single-element standard for IC testing
Supplier: SCP SCIENCE CA
$271.98
Copper ≥99.9% (metals basis), shot 1-10 mm (0.04-0.4 in)
Supplier: Thermo Scientific Chemicals
$106.00
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$156.54 to $571.97