PRESSURE SENSOR BULKHEAD
Catalog #: MFLX00320-XW
Supplier: PENDOTECH
Customers Who Bought This Also Bought
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
$729.52
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$619.49 to $3,535.84
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Argos Technologies
$62.31
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Sodium phosphate dibasic, anhydrous ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
$622.34