Polypropylene flexible liner pump, rubber liner, 1 GPM, 115/230 VAC
Catalog #: MFLX79440-00
Supplier: VANTON PUMP & EQUIPMENT CORP
Specifications
See all product configurations- Catalog No:
- MFLX79440-00
- MFLX79440-30
- Duty cycle continuous:
- Yes
- Yes
- Inlet connection:
- 3/4" hose barb
- 1-1/4" hose barb
- Max. temperature:
- 85 °C (185 °F)
- 85 °C (185 °F)
- Motor hp:
- 1/4
- 1/2
- Outlet connection:
- 3/4" hose barb
- 1" hose barb
- Run dry:
- Yes
- Yes
- Suction lift, dry:
- 1800 mm of H₂O (72" H₂O); 629 mm Hg (5.3" Hg)
- 1800 mm of H₂O (72" H₂O); 629 mm Hg (5.3" Hg)
- Wetted materials:
- Polypropylene, Nordel®
- Polypropylene, Nordel®
- Warranty:
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Unique sealless design permits gentle pumping action that provides little churning or foaming.
- Transfer liquids containing up to 20% soft solids
- Sealless design with secondary containment system prevents potentially hazardous leakage
- Fixed-speed pumps are self-priming, reversible, and can run dry without damage
Polypropylene pumps have a Nordel® rubber liner; PTFE pumps have a Hypalon® rubber liner. Variable-speed models and DC-powered pumps are available — call our Applications Specialists for more information.
- Maximum Flow Rate:3.8 L/min (1 gal./min)
- Description:Polypropylene flexible liner pump, rubber liner, 1 gal./min, 115/230 VAC
- Height:206.4 mm
- Power:115/230 VAC; 60 Hz; 3.5 A
- Wetted materials:Polypropylene, Nordel®
- Duty cycle continuous:Yes
- Inlet connection:3/4" hose barb
- Motor hp:1/4
- Outlet connection:3/4" hose barb
- Run dry:Yes
- Suction lift, dry:1800 mm of H₂O (72" H₂O); 629 mm Hg (5.3" Hg)
- Max. temperature:85 °C (185 °F)
- Warranty:1 year
- Cat. no.:MFLX79440-00
- Length:38.1 cm
- Width:17.78 cm
Customers Who Bought This Also Bought