Pulsafeeder K4VTC1 Service Kit for Solenoid Metering Pump, 21- 44 GPD
Catalog #: MFLX78118-82
Supplier: EQUIP SOLUTIONS
Specifications
See all product configurations- Warranty:
- 3 months
- 3 months
- 3 months
- 3 months
- 3 months
- Currently Viewing
Specifications
About this item
Optimize use of your Pulsafeeder metering pumps.
- Service kits are suitable for use with Pulsafeeder solenoid metering pumps
Replace wearable parts to extend the life of your pump.
- Description:Service Kit for Solenoid Metering Pump, 21- 44 GPD
- Warranty:3 months
- Cat. no.:MFLX78118-82
Customers Who Bought This Also Bought