Masterflex® I/P® Easy-Load® Rotor and Bearing Support Assembly
Catalog #: MFLX77601-68
Supplier: Avantor Fluid Handling
Specifications
See all product configurations- Catalog No:
- MFLX77601-68
- MFLX77602-28
- MFLX77602-38
- Pump series:
- I/P
- I/P
- I/P
- Warranty:
- 3 months
- 3 months
- Currently Viewing
Specifications
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
- Suitable for use with I/P® Easy-Load® pump heads
Replacement parts to keep your pump head operating even longer.
- Pump series:I/P
- Description:I/P Rotor/Bearing Support Assembly for Easy-Load 77601-60
- Cat. no.:MFLX77601-68
- Rotor Material:Polyphenylene sulfide (PPS)
Customers Who Bought This Also Bought