Abaque 104884 Lubricant - 1 Gallon
Catalog #: MFLX75810-89
Supplier: NEPTUNE MIXER CO
CE Compliant
Specifications
See all product configurationsSpecifications
About this item
Replacement lubricant for the liquid end coming in four different volumnes.
- Original lubricant designed to help extend tubing life and to dissipate heat build up.
Optimize performance of your Abaque™ positive displacement pumps.
- Description:Abaque™ lubricant for positive displacement pumps, 3785 ml
- Warranty:1 year
- Cat. no.:MFLX75810-89
Customers Who Bought This Also Bought