Walchem Z20PC-PK Service Kit For 74126-20, -25 And 74127-20, -25
Catalog #: MFLX74126-28
Supplier: EQUIP SOLUTIONS
Specifications
See all product configurationsSpecifications
About this item
Extend the life of your pump.
- Easy and quick maintenance
Easy to install, these service kits avoid costly downtime of your pump.
Delivery: Includes diaphragm and retainer, valve guides, valve seats, valve balls, valve gaskets, and o-rings.
- Description:Service Kit For 74126-20. -25 And 74127-20. -25
- Warranty:2 years
- Cat. no.:MFLX74126-28
Customers Who Bought This Also Bought