Pulsafeeder Solenoid Metering Pump Five-Function Valve, PVDF, 150 psi, 3/8" OD
Catalog #: MFLX74113-86
Supplier: EQUIP SOLUTIONS
Specifications
See all product configurations- Warranty:
- 1 year
- 3 months
- 3 months
- 1 year
- Currently Viewing
Specifications
About this item
Optimize performance of your Pulsafeeder Solenoid Metering Pump.
- Choose from a wide range of replacement parts and accessories
Complete and customize a pump system for any application with a variety of valves and mounting options. Service kits are also available to make routine maintenance easy.
- Description:PVDF five-function valve, compression fitting, 3/8" tubing connectors, 150 psi max
- Warranty:1 year
- Cat. no.:MFLX74113-86
Customers Who Bought This Also Bought