Universal Drum Wrench, Dual-Ended, Non-Sparking Brass
Catalog #: MFLX70607-91
Supplier: Action Pump
Specifications
See all product configurations- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- None
- 1 year
- Currently Viewing
Specifications
About this item
Remove or tighten plugs on most industrial drums.
- Fiberglass-reinforced nylon wrenches are lightweight and easy to carry
- Use the nonsparking brass alloy models for opening drums containing flammables
- More corrosion resistant than steel versions
Use the round end for outside splines of bung caps. Use the long end for internal splines of bung caps.
Round end for outside splines of bung caps. Long end for internal splines of bung caps.
- Description:Universal Drum Wrench, Dual-Ended, Non-Sparking Brass
- Warranty:1 year
- Cat. no.:MFLX70607-91
Customers Who Bought This Also Bought