Masterflex® Drum Pump Strainer, Polypropylene; 1/Each
Catalog #: MFLX70604-86
Supplier: VWR International
Specifications
See all product configurationsSpecifications
About this item
Extend the life of your drum pump!
- Optimize performance of your Masterflex® Drum and Container Pumps
Replacement parts are compatible with Masterflex® Complete Drum and Container Pumps, 70613-series.
- Description:Strainer for Drum Pumps, PP
- Material:PP
- Warranty:4 months
- Cat. no.:MFLX70604-86
Customers Who Bought This Also Bought