GF Signet 3-9900 Batch Module for Flow Transmitter 56560-12
Catalog #: MFLX56560-13
Supplier: BURT PROCESS EQUIPMENT
Specifications
See all product configurations- Control type:
- On/Off
- Warranty:
- See message
- 2 years
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- Currently Viewing
Specifications
About this item
Optimize performance of your GF+ Signet Transmitter/Controller.
- Choose from a wide range of replacement parts and accessories
Use modules, sensors, and RC filters to expand the controller's capabilities. Adapt your controller to your existing system with a variety of mounting options.
- Control type:On/Off
- Description:Batch Module for Flow Transmitter 56560-12
- Warranty:See message
- Cat. no.:MFLX56560-13
Customers Who Bought This Also Bought