McMillan Flow Power supply/output cable (0 to 5 VDC), 115 VAC.
Catalog #: MFLX32707-50
Supplier: MC MILLAN CO
Specifications
See all product configurationsSpecifications
About this item
Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.
- Description:Power supply/output cable (0 to 5 VDC), 115 VAC.
- Warranty:1 year
- Cat. no.:MFLX32707-50
Customers Who Bought This Also Bought