Order Entry
ContactUsLinkComponent
Hedland H605B-010 Water Flowmeter, 1.0-10 GPM, Brass, 1/2" NPT(F)
Hedland H605B-010 Water Flowmeter, 1.0-10 GPM, Brass, 1/2" NPT(F)
Catalog #: MFLX32509-75
Supplier:  BADGER METER INC
Hedland H605B-010 Water Flowmeter, 1.0-10 GPM, Brass, 1/2" NPT(F)
Hedland H605B-010 Water Flowmeter, 1.0-10 GPM, Brass, 1/2" NPT(F)
Catalog #: MFLX32509-75
Supplier:  BADGER METER INC
  • Max. operating temperature:
  • 115.5 °C (240 °F)
  • 115.5 °C (240 °F)
  • 115.5 °C (240 °F)
  • 115.5 °C (240 °F)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Currently Viewing

Specifications

About this item

Easy to read scale.

  • Direct reading
  • Install in any position
  • 360º Rotatable Guard/Scale
  • Easier ot Read Linear Scale
  • No Flow Straighteners or Special Piping Required
  • Insensitive to Shock and Vibration
  • Good Viscosity Stability
  • Temperature up to 240ºF
  • Accuracy ±2% Full Scale
  • Repeatability ±1%
  • Special Scales Available

Ideal for applications that are sensitive to shock and vibration.

  • Description:
    Water Flowmeter. 1.0-10 gal./min. Brass. 1/2" NPT(F)
  • Max. operating temperature:
    115.5 °C (240 °F)
  • Warranty:
    1 year
  • Cat. no.:
    MFLX32509-75
Customers Who Bought This Also Bought