Masterflex® Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK
Catalog #: MFLX30600-43
Supplier: VWR International
Specifications
See all product configurations- Wetted materials:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Warranty:
- Out of box
- Out of box
- Out of box
- Currently Viewing
Specifications
About this item
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
- Description:Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Cat. no.:MFLX30600-43
- Flow Pattern:5 ports, 360° rotation
- Inlet Port:Female luer
- Outlet Port:Female luer
Customers Who Bought This Also Bought