Masterflex® Smart Gasket® Hole Plug for System, Stainless Steel; 1/EA
Catalog #: MFLX30545-98
Supplier: VWR International
Specifications
See all product configurations- Catalog No:
- MFLX30545-98
- MFLX30545-99
- Connection type:
- Sanitary
- Sanitary
- Warranty:
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Optimize performance of your Masterflex® Smart Gasket® Sanitary Gasket system.
- Choose from a wide range of replacement parts and accessories
Smart Gaskets allow you to measure temperature, take samples, or dose chemicals in your process line. Selected accessories include thermocouple probes and wire, port plugs for the Smart Gasket, and a sampling tube.
- Connection type:Sanitary
- Description:Hole Plug for System, Stainless Steel; 1/EA
- Warranty:1 year
- Cat. no.:MFLX30545-98
- Fitting Material:Stainless Steel
Customers Who Bought This Also Bought