Manual Purge Kit for Liquid/Solids In-Line Separators
Catalog #: MFLX29995-60
Supplier: VWR International
Specifications
See all product configurations- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Optimize performance of your Liquid-Solids Separator.
- Keep your Separators running properly with these kits
Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
- Description:Manual Purge Kit for Liquid/Solids In-Line Separators
- Warranty:1 year
- Compatible with:29995 series liquid-solid separators/concentrators
- Cat. no.:MFLX29995-60
Customers Who Bought This Also Bought