Mounting Bracket Kit for In-Line Separators, 1/2" to 3/4" Inlet/Outlet
Catalog #: MFLX29995-50
Supplier: VWR International
Specifications
See all product configurations- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Optimize performance of your Liquid-Solids Separator.
- Keep your Separators running properly with these kits
Mounting bracket kits are designed to support the separator at a near-upright position.
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
- Description:Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1/2" to 3/4" Inlet/Outlet
- Warranty:1 year
- Compatible with:29995-00, 29995-02, 29995-04
- Cat. no.:MFLX29995-50
Customers Who Bought This Also Bought