Flowline LU10-2305 Ultrasonic Level Switch-PFA, Short, 3/4" NPT
Catalog #: MFLX23002-16
Supplier: FLOWLINE
IP65 washdown
Specifications
See all product configurations- Catalog No:
- MFLX23002-16
- MFLX23002-14
- Accuracy of depth range:
- Min. process temperature:
- -40 °F
- -40 °F
- Max. process temperature:
- 80 °C
- 80 °C
- Max. pressure:
- 150 psi
- 150 psi
- Relay output:
- SPST relay rated for 60 VA
- SPST relay rated for 60 VA
- Wetted materials:
- PFA
- Polypropylene
- Warranty:
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Compatible with Smart Trak and Switch-Pak installation fittings.
- CSA approved intrinsically safe for use in hazardous environments
- 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.
- Max. pressure:150 psi
- Description:Ultrasonic Level Switch-PFA, Short, 3/4" NPT
- Power:12 - 36 VDC
- Wetted materials:PFA
- Max. process temperature:80 °C
- Min. process temperature:-40 °F
- Relay output:SPST relay rated for 60 VA
- Warranty:1 year
- Cat. no.:MFLX23002-16
Customers Who Bought This Also Bought