ContactUsLinkComponent
GF Signet 161.514.616 Type 514 PVC Manual Diaphragm Valve, 1-1/2"
GF Signet 161.514.616 Type 514 PVC Manual Diaphragm Valve, 1-1/2"
Catalog # MFLX07390-54
undefined
GF Signet 161.514.616 Type 514 PVC Manual Diaphragm Valve, 1-1/2"
Catalog # MFLX07390-54
Supplier Number:  161.514.616
  • Max. temperature:
  • 65.5 °C (150 °F)
  • 65.5 °C (150 °F)
  • 65.5 °C (150 °F)
  • Wetted materials:
  • PVC, EPDM
  • PVC, EPDM
  • PVC, EPDM
  • Working pressure:
  • 150 psi
  • 150 psi
  • 150 psi
  • Warranty:
  • None
  • None
  • None
  • Currently Viewing
Specifications
About this item
Optimized valve body provides double the CV values.
  • Two-color positition indicator on handwheel
  • Elimination of bolts removes corrosive weak points
  • Mount vertically or horizontally
  • PVC body and EPDM diaphragm
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150 °F (65 °C) and feature true union socket connections.
  • Connection Type:
    True union socket
  • Description:
    Type 514 PVC Manual Diaphragm Valve, 1-1/2"
  • Environmentally Preferable:
  • Wetted materials:
    PVC, EPDM
  • Max. temperature:
    65.5 °C (150 °F)
  • Warranty:
    None
  • Working pressure:
    150 psi
  • Cat. no.:
    MFLX07390-54
  • Cv:
    85.3
  • Inlet Port:
    1-½" true union socket
  • Outlet Port:
    1 - ½" true union socket
Frequently Bought Together