Masterflex® L/S® Easy-Load® Pump Head Mounting Hardware, Four Pump Heads, Stainless Steel; 2/Set
Catalog #: MFLX07013-09
Supplier: Avantor Fluid Handling
About this item
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
- Description:L/S® Easy-Load® Pump Head Mounting Hardware, Four Pump Heads, Stainless Steel; 2/Set. 2 years warranty
- Material:Stainless steel
- Compatible with:Masterflex L/S® Easy-Load® Pump head
- Cat. no.:MFLX07013-09
- Pump Series:L/S
Customers Who Bought This Also Bought