Masterflex® Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Catalog #: MFLX06833-00
Supplier: VWR International
Specifications
See all product configurations- Catalog No:
- MFLX06833-00
- MFLX06833-01
- Housing material:
- Polyoxymethylene (POM) Acetal
- Polyoxymethylene (POM) Acetal
- Pinch valve tube size:
- 1/8" to 1/2" OD
- 1/16" to 1/4" OD
- Warranty:
- 3 months
- None
- Currently Viewing
Specifications
About this item
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
- Housing material:Polyoxymethylene (POM) Acetal
- Description:Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
- Pinch valve tube size:1/8" to 1/2" OD
- Warranty:3 months
- Cat. no.:MFLX06833-00
Customers Who Bought This Also Bought