Order Entry
ContactUsLinkComponent
Stainless Steel (SS) Hose Clamps, 7/32" x 5/8"; 10/Pk
Stainless Steel (SS) Hose Clamps, 7/32" x 5/8"; 10/Pk
Catalog #: MFLX06403-11
Supplier:  VWR International
Stainless Steel (SS) Hose Clamps, 7/32" x 5/8"; 10/Pk
Stainless Steel (SS) Hose Clamps, 7/32" x 5/8"; 10/Pk
Catalog #: MFLX06403-11
Supplier:  VWR International

About this item

Ideal for securely clamping tubes in place.

  • Resists corrosion

The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.

  • Max. I.D.:
    15.875 mm (0.625")
  • Description:
    Stainless Steel (SS) Hose Clamps, 7/32" x 5/8"; 10/Pk
  • Material:
    Stainless steel
  • Cat. no.:
    MFLX06403-11
  • Max. Tubing O.D.:
    15.875 mm (0.625")
  • Min. I.D.:
    5.55625 mm (0.21875")
Customers Who Bought This Also Bought