Order Entry
ContactUsLinkComponent
GF Signet Installation Tee Fitting, CPVC 80, 1" Pipe ID
GF Signet Installation Tee Fitting, CPVC 80, 1" Pipe ID
Catalog #: MFLX05620-33
GF Signet Installation Tee Fitting, CPVC 80, 1" Pipe ID
GF Signet Installation Tee Fitting, CPVC 80, 1" Pipe ID
Catalog #: MFLX05620-33
  • Connection type:
  • 1" ID Slip Fit
  • 1/2" ID Slip Fit
  • 1" ID Slip Fit
  • 1-1/2" ID Slip Fit
  • 2" ID Slip Fit
  • 4" ID Slip Fit
  • Warranty:
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor
  • 1 year part and 2 years labor
  • Currently Viewing

Specifications

About this item

Easily install your flowmeters.

  • Constructed of durable CPVC/PVC 80

PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads) sensors.

  • Description:
    Installation Tee Fitting. CPVC 80. 1" Pipe ID
  • Warranty:
    1 year part and 2 years labor
  • Connection type:
    1" ID Slip Fit
  • Cat. no.:
    MFLX05620-33
  • Fitting Material:
    CPVC 80
Customers Who Bought This Also Bought