PVC Pressure Regulator Valve, 1/2" NPT(F)
Catalog #: MFLX01350-82
Supplier: VWR International
Specifications
See all product configurationsSpecifications
About this item
Easily control your outlet pressure.
- Choose between PVC or polypropylene (PP) construction with Viton® diaphragm
Valves feature a diaphragm made of Viton®. Use to control your outlet pressure between 5 and 50 psig; up to 150 psig inlet. Ideal for your liquid applications.
- Connection type:NPT(F)
- Description:PVC Pressure Regulator Valve, 1/2" NPT(F)
- Wetted materials:PVC
- Warranty:1 year
- Cat. no.:MFLX01350-82
- Inlet Port:½" NPT(F)
- Outlet Port:½" NPT(F)
Customers Who Bought This Also Bought