TUBING TYGON 2475 1/16"ID 50'
Catalog #: MFLX00364-LY
Supplier: Saint Gobain Life Sciences
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$389.38 to $1,030.72
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Argos Technologies
$62.31
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$72.65 to $846.50
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50