GEAR PUMPHEAD 0.91 ML/REV
Catalog #: MFLX00313-SX
Supplier: MICROPUMP INC
Customers Who Bought This Also Bought
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$214.10 to $1,039.32
Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
$428.18
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, sterile
Supplier: Teknova
$386.34
Rubidium hydroxide 50% (w/w) in aqueous solution , 99.6% (metals basis)
Supplier: Thermo Scientific Chemicals
$212.00
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80