MAGNET HUB ASSEMBLY
Catalog #: MFLX00104-DZ
Supplier: MICROPUMP INC
Customers Who Bought This Also Bought
Hydrogen peroxide 6% in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
$133.13 to $1,428.48
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
$2,699.72
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Tryptic Soy Agar with Polysorbate 80 and Lecithin, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
$530.43
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61
Sabdex Agar with Chloramphenicol, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$214.45