Cole-Parmer® Grinding Balls
Catalog #: 76533-064
Supplier: SPEX
Specifications
- Description:Grinding balls, pack of 100
- Material:Stainless steel
- Size:6 mm (¹/₄")
- Cat. no.:76533-064
- For Use With:Spex® SamplePrep homogenizers, grinders, and mills
Specifications
About this item
Crush, grind, and mill various materials.
- Use grinding balls to grind samples in Cole-Parmer® homogenizers and grinders
Replace vial set grinding balls to ensure optimal grinding potential.
Customers Who Bought This Also Bought