Order Entry
ContactUsLinkComponent
FLASK VOLUMETRIC CLASS A 5ML
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, United Scientific Supplies
Catalog #: 76531-850
FLASK VOLUMETRIC CLASS A 5ML
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, United Scientific Supplies
Catalog #: 76531-850

Specifications

  • Capacity:
    5 ml
  • Cat. no.:
    76531-850
  • Stopper No.:
    9
  • Tolerance:
    ±0.02 ml

Specifications

About this item

Durable, machine-blown body with heavy beaded tubing neck.

  • Sharp graduation line and large white block lettering

A ground glass stopper is included.

Certifications: Complies with ASTM E288 Class A specifications; individually serialized and certified.
Customers Who Bought This Also Bought