Order Entry
ContactUsLinkComponent
FLASK VOLUMETRIC CLASS A QR CODED 1000ML
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, QR, United Scientific Supplies
Catalog #: 76531-824
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, QR, United Scientific Supplies
Catalog #: 76531-824

Specifications

  • Capacity:
    1000 ml
  • Cat. no.:
    76531-824
  • Stopper No.:
    22
  • Tolerance:
    ±0.30 ml

Specifications

About this item

Durable, machine-blown body with heavy beaded tubing neck.

  • Sharp graduation line and large white block lettering

A printed QR code allows for easy access to the calibration certificate from a mobile device or computer. A paper certificate is also included.

Certifications: Complies with ASTM E288 Class A specifications; individually serialized and certified.
Ordering information: A ground glass stopper is included.
Customers Who Bought This Also Bought