Ward's® Live Waxworms (Galleria mellonella)
Catalog #: 470336-812
Supplier: DMF BAIT COMPANY
Specifications
- Description:Pack of 250
- Cat. no.:470336-812
Specifications
About this item
Wax worms are a hardy larvae that are easy to handle.
- Chemical-free, safe for feeding to pets
- Healthy for many reptiles and fish
- These creamy white worms have soft, grublike bodies and are dry to the touch
- Refrigeration optional
Wax worms do not need moisture, and you do not need to feed them as in their larva state they live off their own body fat. You can keep them at room temperature, or around 45 to 65 °F for maximum shelf life.
Delivery: These products contain living materials and will ship via overnight or 2 nd day delivery. Please specify a delivery date during checkout.
Packaging: Packed in cups with wood chips.
Customers Who Bought This Also Bought