Order Entry
ContactUsLinkComponent
KIT GENFIND V3 REAGENT LIQUID 384 PREPS
GenFind V3 Reagent Kits, Beckman Coulter
Catalog #: CA76409-761
Supplier:  Beckman Coulter
CAS Number:  
KIT GENFIND V3 REAGENT LIQUID 384 PREPS
GenFind V3 Reagent Kits, Beckman Coulter
Catalog #: CA76409-761
Supplier:  Beckman Coulter
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Liquid
  • Sample type:
    Blood
  • Description:
    GenFind V3 reagent kit
  • Size:
    384 preparations
  • Cat. no.:
    CA76409-761
  • Application:
    Genomic DNA extraction from whole blood

Specifications

About this item

This prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit is uses Solid Phase Reversible Immobilization (SPRI) paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications such as PCR.

  • Automated available
Customers Who Bought This Also Bought