VWR Probe Holder, Temperature, Ult25Neu Probe
Catalog #: 76530-430
Supplier: Stirling Ultracold
Customers Who Bought This Also Bought
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Supplier: Air-Tite
$36.99 to $141.80
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$279.98 to $1,119.95
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Magnesium chloride hexahydrate 99.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$137.92 to $1,374.20
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80