ContactUsLinkComponent
P-Cadherin Monoclonal Antibody, Clone: 12H6, Host Species: Mouse, Reactivity: Human, Isotype: IgG1, K, Conjugate: Purified, Immunogen: Recombinant full-length Human P-cadherin peptide, Concentration: 0.5 mg/mL, Application: WB, IP, Size: 25uG
Catalog #: CA76523-092
Supplier:  Biolegend
P-Cadherin Monoclonal Antibody, Clone: 12H6, Host Species: Mouse, Reactivity: Human, Isotype: IgG1, K, Conjugate: Purified, Immunogen: Recombinant full-length Human P-cadherin peptide, Concentration: 0.5 mg/mL, Application: WB, IP, Size: 25uG
Catalog #: CA76523-092
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.