ContactUsLinkComponent
GMP Recombinant Human TPO (carrier-free), Source: Human TPO, amino acids Ser22-Gly353, was expressed in 293E cells, C-terminal contains an 8His-(TG8HGGQ)-tag. Molecular Mass: 345 amino acid recombinant protein has a predicted molecular mass of approximately 37 kD. Purity > 95%, Size: 25uG
Catalog #: CA76522-926
Supplier:  Biolegend
GMP Recombinant Human TPO (carrier-free), Source: Human TPO, amino acids Ser22-Gly353, was expressed in 293E cells, C-terminal contains an 8His-(TG8HGGQ)-tag. Molecular Mass: 345 amino acid recombinant protein has a predicted molecular mass of approximately 37 kD. Purity > 95%, Size: 25uG
Catalog #: CA76522-926
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.