ContactUsLinkComponent
CD274 (B7-H1, PD-L1) Monoclonal Antibody, Clone: 29E.2A3, Host Species: Mouse, Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Isotype: IgG2b, K, Conjugate: PE/Fire 810, Immunogen: Full length human PD-L1, Application: Flow cytometry, Size: 25 tests
Catalog #: CA76522-858
Supplier:  Biolegend
CD274 (B7-H1, PD-L1) Monoclonal Antibody, Clone: 29E.2A3, Host Species: Mouse, Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Isotype: IgG2b, K, Conjugate: PE/Fire 810, Immunogen: Full length human PD-L1, Application: Flow cytometry, Size: 25 tests
Catalog #: CA76522-858
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.