GMP Recombinant Human IL-4 (carrier-free), Source: Human IL-4, amino acids His25-Ser153 was expressed in E. Coli. Molecular Mass: 130 amino acid recombinant protein has a predicted molecular mass of approximately 15.1 kD. The N-terminal amino acid is Met. Purity > 95%, Size: 100uG
Catalog #: CA76522-820
Supplier: Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.