ContactUsLinkComponent
IgG2a, K Isotype Ctrl Monoclonal Antibody, Clone: MOPC-173, Host Species: Mouse, Conjugate: Spark YG 593, Formulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA), Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-750
Supplier:  Biolegend
IgG2a, K Isotype Ctrl Monoclonal Antibody, Clone: MOPC-173, Host Species: Mouse, Conjugate: Spark YG 593, Formulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA), Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-750
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.