ContactUsLinkComponent
CD326 (Ep-CAM) Monoclonal Antibody, Clone: 10D6, Host Species: Mouse, Reactivity: Rat, Isotype: IgG1, K, Conjugate: Alexa Fluor* 594, Concentration: 5.0 - 10.0 ug/ml, Application: IHC-F, Size: 25uG
Catalog #: CA76522-724
Supplier:  Biolegend
CD326 (Ep-CAM) Monoclonal Antibody, Clone: 10D6, Host Species: Mouse, Reactivity: Rat, Isotype: IgG1, K, Conjugate: Alexa Fluor* 594, Concentration: 5.0 - 10.0 ug/ml, Application: IHC-F, Size: 25uG
Catalog #: CA76522-724
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.