ContactUsLinkComponent
MOB1A Monoclonal Antibody, Clone: W20004B, Host Species: Rat, Reactivity: Human, Mouse, Isotype: IgG2b, K, Conjugate: Purified, Immunogen: Recombinant human MOB1A protein, Concentration: 0.5 mg/mL, Application: WB, IP, Size: 25uG
Catalog #: CA76522-720
Supplier:  Biolegend
MOB1A Monoclonal Antibody, Clone: W20004B, Host Species: Rat, Reactivity: Human, Mouse, Isotype: IgG2b, K, Conjugate: Purified, Immunogen: Recombinant human MOB1A protein, Concentration: 0.5 mg/mL, Application: WB, IP, Size: 25uG
Catalog #: CA76522-720
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.